Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNAP29 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11363R-HRP
BOSSBS-11363R-HRPEA 900 EUR InStock
BOSSBS-11363R-HRP
Anti-SNAP29 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
SNAP 29 is a 258 amino acid protein that localizes to the membrane and the cytoplasm, as well as to the cell junction, and contains one t-SNARE coiled-coil homology domain. Expressed in liver, heart, brain, kidney, placenta, lung, spleen, pancreas and skeletal muscle, SNAP 29 binds tightly to Syntaxins and, via this binding, is involved in membrane trafficking events. Defects in the gene encoding SNAP 29 are the cause of CEDNIK syndrome, a neurocutaneous syndrome that is associated with cerebral dysgenesis, neuropathy, ichthyosis and palmoplantar keratoderma. The gene encoding SNAP 29 maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SNAP29
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge