Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SV2C Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11366R-A350
BOSSBS-11366R-A350EA 893 EUR InStock
BOSSBS-11366R-A350
Anti-SV2C Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
In all vertebrates, SV2 proteins are abundant, hydrophobic, membrane glycoproteins that are expressed as two major isoforms, SV2A and SV2B, and one minor isoform, SV2C. SV2 proteins are differentially expressed in the brain and are present on all synaptic vesicles, independent of transmitter type. SV2A is abundantly expressed in the subcortex, specifically in the synaptic vesicles of all presynaptic nerve terminals, and also in most neuroendocrine secretory granules. SV2B displays a more restricted pattern of expression in that it is only present on a small subset of synapses in the hippocampus and cortex. SV2A and SV2B are funtionally redundant and are required for maintaining normal brain function in vertebrates. SV2A and SV2B mediate synaptic transmission by regulating cytoplasmic Ca2+ levels in the nerve terminal during repetitive stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SV2C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge