Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-STX12 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11378R-A350
BOSSBS-11378R-A350EA 876 EUR InStock
BOSSBS-11378R-A350
Anti-STX12 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Syntaxin 13 is an integral membrane protein that belongs to the t-SNARE family, a group of proteins involved in protein transport. Confocal immunofluoresence and electron microscopy studies have shown that syntaxin 13 is primarily localized to tubular early and recycling endosomes, where it colocalizes with transferrin receptor, and it is also localized in endosomal vacuoles. Syntaxin 13 has been found to be expressed in all tissues, with higher levels of the protein found in brain, lung, spleen, thymus and testes. Immunoprecipitation studies show that syntaxin 13 complexes with beta-SNAP, VAMP2/3, and SNAP25. The binding of this complex to alpha-SNAP and NSF is terminated in the presence of ATP. These results suggest that syntaxin 13 is a SNARE protein which mediates the recycling protein flow through tubulo vesicular recycling endosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STX12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge