Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AASDHPPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11390R-A488
BOSSBS-11390R-A488EA 893 EUR InStock
BOSSBS-11390R-A488
Anti-AASDHPPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
AASDHPPT (aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase), also known as LYS2, LYS5 or CGI-80, is a 309 amino acid protein that localizes to the cytoplasm and belongs to the P-Pant transferase superfamily. Expressed in testis, liver, kidney, heart, brain, placenta and skeletal muscle, AASDHPPT exists as a monomer that functions to catalyze the phosphopantetheine-dependent post-translational modification of target proteins, effectively transferring a 4'-phosphopantetheine moiety from coenzyme A (CoA) to a serine residue of an acceptor protein. AASDHPPT is subject to DNA damage-dependent phosphorylation, probably by ATM or ATR. The gene encoding AASDHPPT maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AASDHPPT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge