Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SYT11 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11399R-CY7
BOSSBS-11399R-CY7EA 900 EUR InStock
BOSSBS-11399R-CY7
Anti-SYT11 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Synaptotagmins are a large family of synaptic vesicle type III integral membrane proteins that function as regulators of both exocytosis and endocytosis and are involved in neurotransmitter secretion from small secretory vesicles. Synaptotagmin XI, also known as SYT11 (Synaptotagmin-11), is a 431 amino acid protein that localizes to the membrane and is expressed ubiquitously with highest expression in brain and lung. Like other Synaptotagmin proteins, Synaptotagmin XI is involved in the calcium-dependent exocytosis of secretory vesicles and is thought to act as a calcium sensor during vesicular trafficking. Synaptotagmin XI contains two C2 domains through which it can bind either three calcium ions or the zinc-finger protein Parkin (a juvenile Parkinson’s disease gene product), the latter of which causes the polyubiquitination and subsequent degradation of Synaptotagmin XI by the proteasome complex. Defects in the gene encoding Synaptotagmin XI are implicated in a number of neurological disorders, including schizophrenia and Parkinson’s disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYT11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge