Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AKR1C3/DD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11401R-A750
BOSSBS-11401R-A750EA 890 EUR InStock
BOSSBS-11401R-A750
Anti-AKR1C3/DD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DD3 is a unique enzyme that can specifically catalyse the dehydrogenation of trans-benzenedihydrodiol and trans-naphthalenedihydrodiol.Human liver contains isoforms of dihydrodiol dehydrogenase (DD1, DD2, DD3 and DD4), which belong to the aldo-oxo reductase/aldo-keto reductase (AKR) superfamily, have 20Alpha- or 3Alpha-hydroxysteroid dehydrogenase (HSD) activity. DD1 is also designated AKR1C1, DDH or DDH1 while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3Alpha-HSD are alternate designations for DD3, while DD4 also can be called AKR1C4, CD or CHDR. DD1 and DD2 are 20Alpha-HSDs, whereas DD3 and DD4 are the 3Alpha-HSDs. The multiple human cytosolic dihydrodiol dehydrogenases are involved in the metabolism of xenobiotics, such as polycyclic aromatic hydrocarbons, pesticides and steroid hormones, and are responsible for the reduction of ketone-containing drugs by using NADH or NADPH as a cofactor. The 20Alpha-HSD catalyses the reaction of progesterone to the inactive form 20Alpha-hydroxyprogesterone. The 3Alpha-HSD is a cytosolic, monomeric, NADPH-dependent oxidoreductase that reduces 3-keto-5-dihydrosteroids to their tetrahydro products. DD1 and DD2 are ubiquitously expressed, whereas DD4 mRNA is restricted to the liver.

Type: Primary
Antigen: AKR1C3/DD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 161-270/323
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge