Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF461 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11406R-A647
BOSSBS-11406R-A647EA 893 EUR InStock
BOSSBS-11406R-A647
Anti-ZNF461 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
GIOT-1 is a 563 amino acid protein belonging to the Krüppel C2H2-type zinc-finger protein family. Localized to the nucleus, GIOT-1 is widely expressed in tissues, with highest levels in liver, kidney, small intestine, pancreas and thymus. GIOT-1 contains 12 C2H2-type zinc fincers and one KRAB domain. Because the KRAB domain functions as a transcriptional repressor when attached to the template DNA, GIOT-1 is thought to be involved in transcriptional regulation. The gene encoding GIOT-1 is localized to chromosome 19q13.12 and two isoforms of GIOT-1 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF461
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge