Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TSKS Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11413R-A350
BOSSBS-11413R-A350EA 893 EUR InStock
BOSSBS-11413R-A350
Anti-TSKS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
TSKS is a 592 amino acid protein that is highly expressed in human testicular tissue. Low levels of TSKS are detectable in prostate, placenta, fetal liver, thymus, and mammary gland tissues. TSKS is found to be downregulated in cancerous testicular tissue from seminoma, teratocarcinoma, embryonal and Leydig cell tumors concurrently with high expression in neighboring premalignant carcinoma. TSKS protein contains an N-terminal signal peptide, but does not contain a transmembrane region. TSKS has many potential phosphorylation and glycosylation sites and is phosphorylated by soluble recombinant TSSK2 in vitro. It is thought that TSKS likely plays a physiological role in spermatogenesis or spermiogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSKS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge