Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF318 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11414R-A680
BOSSBS-11414R-A680EA 890 EUR InStock
BOSSBS-11414R-A680
Anti-ZNF318 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ZNF318 is a 2279 amino acid endocrine regulatory protein that localizes to the nucleus. Highly expressed in testis, ovaries and kidneys, ZNF318 is a co-repressor of androgen receptor (AR)-mediated transcriptional activation and is thought to regulate transcription during spermatogenesis. ZNF318 interacts with the N-terminal domain of AR and contains two matrin-type zinc fingers. Two isoforms of ZNF318, designated TZF and TZF-L, are produced due to alternative splicing events. Each of these splice variants are thought to have unique roles in transcriptional regulation. While the TZF isoform functions as a repressor of AR-mediated transcriptional activation, the TZF-L isoform is thought to enhance AR-mediated transcriptional activation.

Type: Primary
Antigen: ZNF318
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1077-1125/2279
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge