Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Sauvagine Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11424R-A488
BOSSBS-11424R-A488EA 893 EUR InStock
BOSSBS-11424R-A488
Anti-Sauvagine Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Sauvagine is a 40-residue peptide isolated and characterized from frog skin. It shares significant structural homology with and elicits similar biological responses as corticotropin-releasing factor (CRF). It has high affinity for both the CRF1 and CRF2 receptors (and low affinity for the CRF binding protein), suggesting its potential as a pharmacological tool for the study of these receptor sub-types. Due to the binding to CRF2 receptors and their role in the release of ACTH and beta-endorphin during stress, possible areas of research with this ligand include anxiety, depression and obesity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Sauvagine
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge