Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MED7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11427R-HRP
BOSSBS-11427R-HRPEA 900 EUR InStock
BOSSBS-11427R-HRP
Anti-MED7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
In mammalian cells, transcription is regulated in part by high molecular weight coactivating complexes that mediate signals between transcriptional activators and RNA polymerase (1). These complexes include the SMCC (SRB and MED protein cofactor complex), which consists of various subunits that share homology with several components of the yeast transcriptional mediator complexes, and including the human proteins Srb7, Med6 (also designated DRIP33) and Med7 (also designated DRIP34) (2,3). SMCC associates with the RNAPII (RNA polymerase II) holoenzyme through Srb7 and, in turn, enhances gene-specific activation or repression induced by DNA-binding transcription factors (4). Med6 and Med7, as well as other components of SMCC, associate with coactivator proteins from the TRAP (thyroid hormone receptor-activating protein) complex and DRIP (for vitamin D receptor interacting protein) complex to facilitate steroid receptor dependent transcriptional activation (4,5). Additionally, SMCC associates with PC4 (positive cofactor 4) to repress basal transcription independent of RNAPII activity (6).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: MED7
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge