Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NPSR1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11430R-CY5
BOSSBS-11430R-CY5EA 900 EUR InStock
BOSSBS-11430R-CY5
Anti-NPSR1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR154 (G-protein coupled receptor 154), also known as NPSR1 (neuropeptide S receptor), GPRA (G-protein coupled receptor for asthma susceptibility) or PGR14, is a 371 amino acid protein that is thought to play a role in autocrine or paracrine signaling pathways. Ubiquitously expressed, GPR154 exists as nine alternatively spliced isoforms. Defects in the gene encoding GPR154 is the cause of asthma-related traits type 2 (ASRT2).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPSR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge