Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Ephexin-1/EPHEXIN Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11458R-A555
BOSSBS-11458R-A555EA 893 EUR InStock
BOSSBS-11458R-A555
Anti-Ephexin-1/EPHEXIN Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
EPHEXIN is a 710 amino acid protein that localizes to both the membrane and the cytoplasm and contains one SH3 domain, one PH domain and one DH domain. Expressed at high levels in brain and present at lower levels in lung tissue, EPHEXIN interacts with EphA4 and functions as a guanine nucleotide exchange factor (GEF) that is capable of activating Rho A, Rac 1 and Cdc42 and is thought to play a role in axon guidance and growth cone collapse. EPHEXIN is subject to Src-dependent phosphorylation, an event that increases the GEF activity of EPHEXIN toward Rho A. Human EPHEXIN, which exists as multiple alternatively spliced isoforms, shares a high degree of sequence homology with its mouse counterpart, suggesting a conserved role between species.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ephexin-1/EPHEXIN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge