Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FARP2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11459R-CY7
BOSSBS-11459R-CY7EA 900 EUR InStock
BOSSBS-11459R-CY7
Anti-FARP2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
FARP2 is a 1,545 amino acid protein that contains one FERM domain, one DH domain and two PH domains. It exists as two alternatively spliced isoforms that are abundantly expressed in brain, lung, and testis as well as in embryonic hippocampal and cortical neurons. FARP2 functions as a Rho-guanine nucleotide exchange factor that activates RAC1 and is thought to regulate neurite remodeling of embryonic neurons. Sema3A binding to neuropilin-1 induces the dissociation of FARP2 from plexin-A1, thereby activating FARP2's Rac GEF activity which is critical for repulsion of outgrowing axons and suppression of neuronal adhesion. Downregulation of the FARP2 gene has been implicated in autism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FARP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge