Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BCAS1/NABC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11462R-A750
BOSSBS-11462R-A750EA 890 EUR InStock
BOSSBS-11462R-A750
Anti-BCAS1/NABC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
NaBC1 is a protein found amplified in most breast carcinoma forms. It is expressed primarily as a cytoplasmic, detergent-stable homodimer that has a tendency to interact with DYNLL1 (PIN) and DYNLL2. Breast tumour lines that exhibit 20q13.2 gene amplification express much higher levels of the protein as compared to the levels found in other breast cancer lines that do not overexpress the NaBC1 mRNA. However, this upregulation does not affect growth rate or anchoring abilities of a cell, indicating the oncogenic properties of NaBC1 differ from that of other oncogenes.

Type: Primary
Antigen: BCAS1/NABC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 542-584/584
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge