Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CWC22 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11464R-A488
BOSSBS-11464R-A488EA 893 EUR InStock
BOSSBS-11464R-A488
Anti-CWC22 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Spliceosomes are multi-protein complexes that are composed of snRNPs (small nuclear ribonucleoproteins) and a variety of associated protein factors, all of which work in concert to regulate the splicing of pre-mRNA, a critical step in the post-transcriptional regulation of gene expression. CWC22 (CWC22 spliceosome-associated protein), also known as NCM, fSAPb or EIF4GL, is a 908 amino acid nuclear protein and component of the spliceosome C complex. CWC22 is associated with the spliceosome prior to catalytic steps and remains associated throughout the reaction. Belonging to the CWC22 family, CW22 contains one MI domain and a MIF4G domain. The gene encoding CWC22 maps to human chromosome 2q31.3. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CWC22
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge