Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NAV1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11467R-A555
BOSSBS-11467R-A555EA 893 EUR InStock
BOSSBS-11467R-A555
Anti-NAV1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Neuron navigator 1 is a 1877 amino acid cytoplasmic protein that is involved in neuronal migration. Neuron navigtor 1 is widely expressed at low levels, though highest expression is found in both adult and fetal nervous tissue. Through interaction with tubulin, Neuron navigator 1 associates with a subset of mirotubule plus ends present in the growth cone. Overexpression of Neuron navigator 1 leads to microtubule bundling, whereas a reduction of its levels causes loss of directionality in the migration of pontine cell leading processes. There are seven isoforms of Neuron navigator 1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuron navigator 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge