Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NGBR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11468R-HRP
BOSSBS-11468R-HRPEA 900 EUR InStock
BOSSBS-11468R-HRP
Anti-NGBR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Nogo is an oligodendrocyte-specific member of the Reticulon family and is a component of CNS white matter that inhibits axon outgrowth, induces collapse of growth cones of chick dorsal root ganglion cells, and inhibits the spreading of 3T3 fibroblasts. Nogo is expressed by oligodendrocytes but not by Schwann cells and associates primarily with the endoplasmic reticulum. Nogo exists in three different splice forms, Nogo-A, -B and -C. NgBR (Nogo-B receptor), also known as nuclear undecaprenyl pyrophosphate synthase 1 homolog, is a 293 amino acid single-pass type I membrane protein that acts as a specific receptor for the amino-terminus of Nogo-B. Through this interaction, NgBR is involved in the regulation of vascular remodeling and angiogenesis. NgBR also enhances Niemann-Pick type C2 protein (NPC2) stabilization. Knockdown of NgBR mRNA leads to decreased NPC2 levels, which results in the hallmarks of NPC2 mutation: increased intracellular cholesterol accumulation and a loss of sterol sensing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NGBR/Nogo B receptor
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge