Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SEMA3E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11480R-HRP
BOSSBS-11480R-HRPEA 900 EUR InStock
BOSSBS-11480R-HRP
Anti-SEMA3E Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Semaphorins are a family of cell surface and secreted proteins involved in neural development that are conserved from insects to humans. Members of this family are approximately 750 amino acids in length (including signal sequences) and are defined by a conserved extracellular “semaphorin” domain of approximately 500 amino acids containing 14-16 cysteines, blocks of conserved sequences and no obvious repeats. The transmembrane semaphorins are characterized by an additional 80 amino acid transmembrane domain and an 80-110 amino acid cytoplasmic domain. Secreted and cell-bound semaphorins chemically attract and repel the growth of neural axons, guiding the development of intricate networks of neural tissue. SEMA3E is a secreted semaphorin with 775 amino acids. Mutations in the SEMA3E gene are associated with CHARGE syndrome, a disorder characterized by cranial nerve dysfunction, coloboma of the eye, choanal atresia, inner and external ear abnormalities, cardiac anomalies, genitourinary abnormalities, and growth retardation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SEMA3E
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge