Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TOR1B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11491R-CY7
BOSSBS-11491R-CY7EA 900 EUR InStock
BOSSBS-11491R-CY7
Anti-TOR1B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
TorsinB is a 336 amino acid protein that localizes to the lumen of endoplasmic reticulum and belongs to the Torsin family. Expressed in a variety of tissues with particularly high levels in adult brain, TorsinB is thought to function as a molecular chaperone that assists in the conformational folding of membrane and secreted proteins. Via its ability to regulate protein folding, TorsinB plays a key role in postnatal developmental events and is essential for proper neurological development. TorsinB contains two PKC (protein kinase C) phosphorylations sites, one ATP binding domain and several conserved cysteines and shares 69% amino acid identity with TorsinA, a related family member. Defects in the gene encoding TorsinB are associated with torsion dystonia, an autosomal dominant neurological disorder that is characterized by debilitating muscle contractions throughout the body.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TOR1B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge