Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UNC5B/UNC5H2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11492R-A750
BOSSBS-11492R-A750EA 890 EUR InStock
BOSSBS-11492R-A750
Anti-UNC5B/UNC5H2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The UNC5H family of proteins act as transmembrane receptors for netrin-1 and play a crucial role in axon guidance and migration of neural cells. In fact, UNC5H receptors express widely in cells that migrate, where they bind the G protein G Alpha 1-2 to inhibit G protein Signalling. Additionally, UNC5H receptors induce apoptosis when cleaved by a caspase, producing an intracellular fragment containing a death domain, but this activity is blocked by the binding of netrin-1. The expression of UNC5H receptors is down-regulated in multiple cancers, including colorectal, breast, ovary, uterus, stomach, lung, and kidney cancers. Hence, in the absence of netrin-1, UNC5H receptors act as tumour suppressors by inhibiting anchorage-independent growth and invasion, but mutation of these receptors provides a potential mechanism for tumourigenicity. UNC5H2, also designated unc-5 homolog B or p53-regulated receptor for death and life protein 1 (p53RDL1) is highly expressed in brain with lower levels of expression observed in developing lung, cartilage, kidney and hematopoietic and immune tissues.

Type: Primary
Antigen: UNC5B/UNC5H2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 188-260/945
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge