Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VAX2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11497R-FITC
BOSSBS-11497R-FITCEA 900 EUR InStock
BOSSBS-11497R-FITC
Anti-VAX2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
VAX2 is a 290 amino acid homeobox protein that plays a crucial role in development of the eye, particularly in the specification of the ventral optic vesicle and in establishment of a correct dorsoventral pattern. VAX2 acts as a transcription factor with VAX1 to cooperatively regulate retinal differentiation, neuroepithelial cell proliferation and axial polarization in the retina. Together, VAX1 and VAX2 repress transcription of Pax-6, a strong inducer of retinal development. VAX1 and VAX2 localize to the nucleus and contain one homeobox DNA-binding domain each.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VAX2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge