Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NENF/Neudesin Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11502R
BOSSBS-11502REA 725 EUR InStock
BOSSBS-11502R
Anti-NENF/Neudesin Rabbit Polyclonal Antibody
Antikörper
Neudesin is a 172 amino acid secreted protein that belongs to the cytochrome b5 family and MAPR subfamily. Neudesin possesses neurotrophic activity, which is enhanced by binding to heme, and may contribute to neuronal differentiation and neural cell proliferation. In primary cultured neurons, Neudesin has been observed to activate Akt1 and ERK 1 phosphorylation. Upregulated in immortal cells, Neudesin contains one cytochrome b5 heme-binding domain and is encoded by a gene that maps to human chromosome 1q32.3. Human chromosome 1 spans 260 million base pairs and comprises nearly 8% of the human genome. A large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome, map to chromosome 1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NENF/Neudesin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge