Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BBS10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11512R-A350
BOSSBS-11512R-A350EA 893 EUR InStock
BOSSBS-11512R-A350
Anti-BBS10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Bardet-Biedl syndrome (BBS) is a pleiotropic genetic disorder characterized by obesity, photoreceptor degeneration, polydactyly, hypogenitalism, renal abnormalities, and developmental delay. Other associated clinical findings in BBS patients include diabetes, hypertension, and congenital heart defects. BBS genes map to multiple loci and encode fourteen proteins, BBS1-BBS14. Many BBS genes encode basal body or cilia proteins, suggesting that BBS is a ciliary dysfunction disorder. BBS10 (Bardet-Biedl syndrome 10), also known as chromosome 12 open reading fame 58, C12orf58 or FLJ23560, is a novel 723 amino acid protein belonging to the TCP-1 chaperonin family. BBS10 localizes to the basal body of primary cilium and assists in protein folding upon ATP hydrolysis. Inhibition of BBS10 has been found to impair ciliogenesis, activate the glycogen synthase kinase 3 pathway and cause peroxisome proliferator-activated receptor nuclear accumulation. The gene encoding BBS10 contains two exons and maps to human chromosome 12q21.2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BBS10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge