Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALP Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11526R-CY7
BOSSBS-11526R-CY7EA 900 EUR InStock
BOSSBS-11526R-CY7
Anti-GALP Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The galanin family of proteins are key members for inflammatory processes and cell proliferation, and may function as potential biomarkers for colon cancer. Produced in both neuronal and nonneuronal cells in the skin, members of the galanin family include galanin, galanin-message associated peptide, galanin-like peptide and alarin. GALP, also known as galanin-like peptide, is a 116 amino acid secreted protein belonging to the galanin family. Mainly produced in the arcuate nucleus of the hypothalamus (ARC) and the posterior pituitary, GALP is thought to function in CNS homeostatic processes, including the regulation of gonadotropin-releasing hormone secretion. GALP binds to the G-protein coupled galanin receptors, including GALR1, GALR2 and GALR3, and may also play a role in energy metabolism, with significant implications towards obesity. GALP exits as two alternatively spliced isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALP/Alarin
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge