Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NPW Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11531R
BOSSBS-11531REA 725 EUR InStock
BOSSBS-11531R
Anti-NPW Rabbit Polyclonal Antibody
Antikörper
NPW is a 165 amino acid secreted protein that is cleaved into two chains: neuropeptide W-23 (also designated NPW23 or L8) and neuropeptide W-30 (also known as NPW30 or L8C). Both NPW neuropeptides activate G-protein coupled receptors in the central nervous system to enhance cortisol secretion. Highly expressed in lymphoblastic leukemia, colorectal adenocarcinoma, fetal kidney, trachea and substantia nigra, NPW is also found at low levels in placenta, ovary, testis and uterus. NPW functions in organization of neuroendocrine signals and is also thought to enhance food and water intake as well as stress responses. The gene encoding NPW maps to human chromosome 16p13.3.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NPW
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge