Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CSP1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11541R-CY5
BOSSBS-11541R-CY5EA 900 EUR InStock
BOSSBS-11541R-CY5
Anti-CSP1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DSCR1 protein influences cardiac and nervous system development (1). DSCR1 is abundant in the brain, heart, and skeletal muscle. Overexpression of DSCR1 may play a role in the pathogenesis of Down syndrome, in particular mental retardation and/or cardiac defects. DSCR1 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A. Human DSCR1 maps to gene locus 21q22.1-q22.2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge