Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RCAN3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11542R-HRP
BOSSBS-11542R-HRPEA 900 EUR InStock
BOSSBS-11542R-HRP
Anti-RCAN3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Calcineurin is a Ca(2+) dependent protein phosphatase that is involed in several neuornal functions. Proteins belonging to the RCAN (Regulator of calcineurin) family modulate the activity of Calcineurin through a valine-rich region within their carboxyl tail. RCANs compete with NFATs (Nuclear factor of activated T-cells) for binding to the same docking site in calcineurin, which results in either inhibition or activation of calcineurin activity. Calcipressin-2, also known as RCAN2 and DSCR1L1 (Down syndrome candidate region 1-like 1), is a 197 amino acid protein that is highly expressed in both the cytoplasm and nucleus of neurons and is the only RCAN family member that is found in glial cells. Calcipressin-3, also known as RCAN3 and DSCR1L2 (Down syndrome candidate region 1-like protein 2), is a 241 amino acid protein that potentially is involved in central nervous system development. Calcipressin-3 interacts with cardiac troponin I, suggesting that it may play a role in cardiac contraction events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RCAN3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge