Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXG1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11557R-HRP
BOSSBS-11557R-HRPEA 900 EUR InStock
BOSSBS-11557R-HRP
Anti-FOXG1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The winged-helix transcriptional repressor (WH) BF-1 gene encodes brain factor 1 (BF-1), also known as foxg1, and is essential for the proliferation of progenitor cells in the cerebral cortex and influences regional patterning in the mammalian telencephalon (1–4). WH proteins are a family of putative transcriptional regulators with diverse roles in development, and are characterized by a highly conserved DNA binding structure, the WH domain (1,5,6). BF-1 plays a critical role in the development of the cerebral hemispheres of the brain and targeted disruption of the gene leads to severe defects in the development of telencephalic structures, such as the cerebral cortex and basal ganglia (1). The loss of BF-1 results in an accelerated rate of neuronal differentiation and the shortening of the neurogenetic period in the embryonic cerebral cortex (1,7). BF-1 is expressed by E8.5 in telencephalic progenitors (1). It may also regulate the response of cerebral cortical progenitors to environmental cues (1).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FOXG1/BF-1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge