Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCNF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11558R-FITC
BOSSBS-11558R-FITCEA 900 EUR InStock
BOSSBS-11558R-FITC
Anti-GCNF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Germ cell nuclear factor (GCNF) is an orphan member of the nuclear receptor gene superfamily that influences neurogenesis and germ cell development. GCNF can homodimerize and bind DNA. GCNF regulates paracrine interaction between the oocyte and somatic cells by regulating the expression of BMP-15 and GDF-9, to affect female fertility. GCNF is present in spermatocytes and round spermatids of adult male mouse testis; northern blot and ribonuclease protection assays have shown GCNF is predominant in the testis. The gene expresses three alternatively spliced transcript variants.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCNF
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge