Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRRTM3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11571R-CY3
BOSSBS-11571R-CY3EA 900 EUR InStock
BOSSBS-11571R-CY3
Anti-LRRTM3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic Alpha/Beta horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The LRRTM protein family plays a role in the regulation of various cellular events during nervous system development. Localizing predominantly to the nervous system, LRRTM family members are known to exhibit synaptogenic activity. LRRTM3 (leucine rich repeat transmembrane neuronal 3) is a 581 amino acid single-pass type I membrane protein belonging to the LRRTM family. LRRTM3 is involved in the development and maintenance of the vertebrate nervous system, and contains ten LRR repeats. Expressed in neuronal tissues, LRRTM3 is encoded by a gene that maps to a region of chromosome 10 that has been linked to late-onset Alzheimer disease and elevated plasma Beta-Amyloid. As a result of alternative splicing events, two LRRTM3 isoforms exist.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRTM3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge