Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11580R-A750
BOSSBS-11580R-A750EA 890 EUR InStock
BOSSBS-11580R-A750
Anti-RAI3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
RAI3 is an transmembrane G-protein-coupled receptor that affects many essential biological processes including embryogenesis, cell growth, differentiation, and apoptosis. RAI3 may also be involved in maintaining homeostasis of epithelial cells. Retinoic acid receptors directly regulate RAI3 during its transcription in embryonal carcinoma differentiation. RAI3 expression is upregulated in most tumour cell lines that express mutant p53, suggesting that p53 interacts with the promoter of RAI3 and represses its expression at the beginning of apoptosis. RAI3 is a potential molecular target for diagnosing breast cancer, and selective suppression of signals from RAI3 may have a place in breast cancer treatments.

Type: Primary
Antigen: RAI3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-33/357
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge