Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-1158R-A750
BOSSBS-1158R-A750EA 899 EUR InStock
BOSSBS-1158R-A750
Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Advanced Glycation End products (AGEs) are the result of a chain of chemical reactions after an initial glycation reaction. The intermediate products are known, variously, as Amadori, Schiff base and Maillard products, named after the researchers who first described them. (The litreature is inconsistent in applying these terms. For example, Maillard reaction products are sometimes considered intermediates and sometimes end products.) Side products generated in intermediate steps may be oxidizing agents (such as hydrogen peroxide), or not (such as beta amyloid proteins). 'Glycosylation' is sometimes used for 'glycation' in the litreature, usually as 'non-enzymatic glycosylation. The AGE-modified BSA was produced by reacting BSA with glycolaldehyde under sterile conditions followed by extensive dialysis and purification steps.
Type: Primary
Antigen: AGEs
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge