Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NAT8B Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11592R-A647
BOSSBS-11592R-A647EA 893 EUR InStock
BOSSBS-11592R-A647
Anti-NAT8B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Acetyltransferases and deacetylases are protein groups most often associated with oncogenesis and cell cycle regulation. NAT-8B (N-acetyltransferase 8B), also known as CML2 (camello-like protein 2), is a 227 amino acid single-pass membrane protein that is implicated in gastrulation regulation. A member of the camello family, NAT-8B contains one N-acetyltransferase domain and is encoded by a gene that maps to human chromosome 2p13.2. The NAT-8B gene is susceptible to a nonsense mutation at Serine 16, which leads to a stop codon and subsequently, a non-functional protein that is truncated in length. Similarly, a nonsense mutation at Glutamine 168 is thought to lead to a non-functional protein, as it causes the N-acetyltransferase to become disrupted. Human chromosome 2 consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstré°‰ syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAT8B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge