Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11595R-A555
BOSSBS-11595R-A555EA 893 EUR InStock
BOSSBS-11595R-A555
Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Three highly similar proteins termed NOMO1, NOMO2 and NOMO3, are encoded by a gene mapping to a region of duplication on the p arm of human chromosome 16. All three NOMO proteins share similar functions and have been difficult to characterize individually. NOMO1 (Nodal modulator 1), also known as PM5, is a 1,222 amino acid highly conserved single-pass type I membrane protein expressed in colon tumor tissue and normal colonic mucosa. NOMO proteins are novel antagonists of Nodal signaling which interact with Nicalin to form a Nicalin-NOMO complex, and are rapidly degraded or stabilized by Nicalin. NOMO proteins were once considered candidates for the development of pseudoxanthoma elasticum (PXE), a heritable disorder of connective tissue, as the NOMO genes are located in close proximity to the gene responsible for PXE development (MRP6).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOMO1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge