Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-11595R-A750
BOSSBS-11595R-A750EA 890 EUR InStock
BOSSBS-11595R-A750
Anti-NOMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Three highly similar proteins termed NOMO1, NOMO2 and NOMO3, are encoded by a gene mapping to a region of duplication on the p arm of human chromosome 16. All three NOMO proteins share similar functions and have been difficult to characterize individually. NOMO1 (Nodal modulator 1), also known as PM5, is a 1222 amino acid highly conserved single-pass type I membrane protein expressed in colon tumour tissue and normal colonic mucosa. NOMO proteins are novel antagonists of Nodal Signalling which interact with Nicalin to form a Nicalin-NOMO complex, and are rapidly degraded or stabilized by Nicalin. NOMO proteins were once considered candidates for the development of pseudoxanthoma elasticum (PXE), a heritable disorder of connective tissue, as the NOMO genes are located in close proximity to the gene responsible for PXE development (MRP6).
Type: Primary
Antigen: NOMO1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 701-800/1222
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge