Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Glyt1/SLC6A9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11604R-A680
BOSSBS-11604R-A680EA 890 EUR InStock
BOSSBS-11604R-A680
Anti-Glyt1/SLC6A9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Na⁺/Cl⁻ dependent neurotransmitter transporters are a superfamily of transmembrane proteins that contain 12 membrane spanning regions. Specifically, the highly hydrophobic Na⁺/Cl⁻ dependent glycine transporters (GlyT) are crucial for the termination of neurotransmission at glycinergic synapses. Two different GlyT genes encode GlyT2 and GlyT1, which exists as two isoforms produced by alternative splicing of the same gene located on human chromosome 1p31.3. The GlyT1 gene may be an early marker of neural development and encodes glia-specific transporter proteins. Although GlyT1 and GlyT2 are both expressed in the brain and spinal cord, each shows a unique pattern of expression. GlyT1 is found only in the white matter of the CNS, whereas GlyT2 is found in the gray matter of the CNS as well as in macrophages and mast cells in peripheral tissues. The anatomic distribution of GlyT2 mRNA suggests that glycine may act as a supraspinal neurotransmitter and may function as a chemical messenger outside the CNS.

Type: Primary
Antigen: Glyt1/SLC6A9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 209-285/706
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge