Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GUCA2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11621R-A488
BOSSBS-11621R-A488EA 893 EUR InStock
BOSSBS-11621R-A488
Anti-GUCA2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The family of guanylin regulatory peptides, including guanylin and uroguanylin, are strongly expressed in intestinal mucosa and regulate intestinal fluid secretion during digestion. Guanylins are also involved in acid neutralization and the regulation of membrane-bound guanylate cyclase signaling molecules. Guanylin and uroguanylin are secreted primarily in the stomach, intestine, and colon. Guanylin is also detected in plasma. Guanylin is an endogenous activator of intestinal guanylate cyclase. It stimulates intestinal guanylate cyclase through the same receptor binding region as the heat-stable enterotoxins. Gut enterochromaffin cells synthesize guanylin to be a prohormone of 115 amino acids which is then is processed to the molecular form of 94 amino acids. This 10kDa form is found circulating in the blood.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Guanylin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge