Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PKD1L3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11622R-CY7
BOSSBS-11622R-CY7EA 900 EUR InStock
BOSSBS-11622R-CY7
Anti-PKD1L3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Polycystin-1L3 is a 1,732 amino acid multi-pass membrane protein that contains one PLAT domain, one GPS domain and one C-type lectin domain. Expressed at high levels in placenta and present at lower levels in lung and heart, Polycystin-1L3 is thought to function as an ion-channel regulator that may interact with Polycystin-L and play a role in heteromeric taste channels. The gene encoding Polycystin-1L3 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PKD1L3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge