Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PIRT Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11623R
BOSSBS-11623REA 725 EUR InStock
BOSSBS-11623R
Anti-PIRT Rabbit Polyclonal Antibody
Antikörper
PIRT is a 137 amino acid multi-pass membrane protein. Highly conserved among vertebrates, PIRT consists of two transmembrane domains and one putative C-terminal phosphoinositide-binding domain. Although PIRT is expressed in peripheral nervous system, with highest levels in dorsal root ganglion and trigeminal neurons, and lowest levels in sympathetic and enteric neurons, it is not expressed in spinal cord. PIRT is a required component of the VR1 complex, which positively regulates VR1, a sensor of both noxious heat and capsaicin. Correspondingly, PIRT knockout results in impaired responses to noxious heat and capsaicin exposure, while VR1 remains unaltered. The gene that encodes PIRT maps to human chromosome 17p13.1.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PIRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge