Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-WBP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11625R-CY5
BOSSBS-11625R-CY5EA 900 EUR InStock
BOSSBS-11625R-CY5
Anti-WBP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
WW domain-binding protein 2 (WBP2) is a 261 amino acid protein expressed in most tissues. The WW domain is composed of 38 to 40 semi-conserved amino acids and is shared by various groups of proteins, including structural, regulatory and signaling proteins. The domain mediates protein-protein interactions through the binding of polyproline ligands. WBP2 binds to the WW domain of Yes-associated protein (YAP), WW domain containing E3 ubiquitin protein ligase 1 (AIP5) and WW domain containing E3 ubiquitin protein ligase 2 (AIP2). The gene encoding WBP2 is located on human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes, some of which are involved in tumor suppression and in the pathogenesis of Li-Fraumeni syndrome, early onset breast cancer and a predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WBP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge