Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-OR10A5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11631R-A750
BOSSBS-11631R-A750EA 890 EUR InStock
BOSSBS-11631R-A750
Anti-OR10A5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Olfactory receptors interact with odorant molecules in the nose to initiate a neuronal response that leads to the perception of smell. While they share a seven transmembrane domain structure with many neurotransmitter and hormone receptors, olfactory receptors are responsible for the recognition and transduction of odorant signals. OR10A2 (olfactory receptor 10A2) and OR10A5 (olfactory receptor 10A5) are multi-pass membrane proteins that belong to the G-protein coupled receptor 1 family and are encoded by genes that map to human chromosome 11p15.4. Chromosome 11 houses over 1400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that map to chromosome 11.

Type: Primary
Antigen: OR10A5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-317/317
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge