Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APBB3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11637R-CY3
BOSSBS-11637R-CY3EA 900 EUR InStock
BOSSBS-11637R-CY3
Anti-APBB3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Fe65L2 is a 486 amino acid protein that contains one WW domain and two PID domains. Binding to the intracellular domain of the ∫-Amyloid precursor protein, Fe65L2 is thought to modulate the internalization and, therefore, the accessibility and function of ∫-Amyloid. Via its ability to control the intracellular accumulation of ∫-Amyloid, Fe65L2 is thought to play a role in the pathogenesis of Alzheimer's disease. Fe65L2 exists as four alternatively spliced isoforms designated isoform I, isoform II, isoform III and isoform IV. Fe65L2 interacts with Amyloid-like protein and is encoded by a gene located on human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APBB3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge