Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APH1B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11640R-HRP
BOSSBS-11640R-HRPEA 900 EUR InStock
BOSSBS-11640R-HRP
Anti-APH1B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Anterior pharynx defective 1 (Aph-1) is a polytopic, seven-pass membrane protein that functions as one of the four essential components in the presenilin-Gamma-secretase enzyme complex. This enzyme complex is necessary for the intra-membrane proteolysis of several different membrane proteins, including the beta-Amyloid precursor protein, and is involved in multiple neurodevelopmental signaling pathways. Aph-1b and Aph-1a are splice variants of Aph-1. Aph-1b specifically lacks exon 4, which encodes for the entire fourth transmembrane domain, causing the protein to be destabilized. Deficiency of Aph-1a causes a reduction in Gamma-secretase activity, however deficiency of Aph-1b does not; thus, Aph-1b may execute redundant functions in the cell. Aph-1b expression and Gamma-secretase activity may be implicated in neurodevelopmental disorders, such as schizophrenia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: APH1B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge