Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CALHM1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11642R
BOSSBS-11642REA 725 EUR InStock
BOSSBS-11642R
Anti-CALHM1 Rabbit Polyclonal Antibody
Antikörper
CALHM1 is a 346 amino acid multi-pass endoplasmic reticulum membrane protein that belongs to the FAM26 family. CALHM1 co-localizes with GRP 78 to the endoplasmic reticulum. Predominantly expressed in adult brain, CALHM1 may be a pore-forming ion channel that controls cytosolic Ca2+ permeability and cytosolic Ca2+ concentration in the cell. It is suggested that CALHM1 regulates amyloid precursor protein proteolysis and aggregated amyloid-beta peptides levels in a Ca2+ dependent manner. CALHM1 homomultimerizes and shares strong sequence similarities with the selectivity filter of the NMDA receptor, which generates a large Ca2+ conductance across the plasma membrane. CALHM1 may be a potential factor involved in the pathogenesis of Alzheimer's disease.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CALHM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge