Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLSTN1/Calsyntenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11644R-A750
BOSSBS-11644R-A750EA 890 EUR InStock
BOSSBS-11644R-A750
Anti-CLSTN1/Calsyntenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Members of the calsyntenin protein family are localised to the post-synaptic membrane of exicitatory central nervous system (CNS) synapses. Calsyntenin-1, also known as CSTN1, PIK3CD, Alzheimer-related cadherin-like protein, non-classical cadherin XB31alpha, KIAA0911, ALC-ALPHA, alcalpha1, alcalpha2 or FLJ32258, is a 981 amino acid single-pass type I membrane protein that localizes to the membrane of endoplasmic reticulum, Golgi apparatus, cell projections and postsynaptic cells. Expressed in brain, calsyntenin-1 is also found at lower levels in placenta, skeletal muscle, heart and kidney. Calsyntenin-1 binds synaptic Ca²⁺ with its cytoplasmic domain and plays a role in extracellular proteolysis. Calsyntenin-1 is also known to form a complex with X11 Beta and APP to suppress the metabolic cleavage of APP, and docks vesicular cargo to KLC1. Calsyntenin-1 may be related to the development or progression of Alzheimer's disease, and two calsyntenin-1 isoforms are produced as a result of alternative splicing events.

Type: Primary
Antigen: CLSTN1/Calsyntenin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/981
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge