Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Humanin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11646R-A680
BOSSBS-11646R-A680EA 890 EUR InStock
BOSSBS-11646R-A680
Anti-Humanin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Humanin, an endogenous anti-apoptotic peptide against Alzheimer disease-related insults, consists of 24 amino acids. The secreted protein is a neuroprotective factor against death induced by several different types of Alzheimer's disease genes. Humanin protects neuronal cells from damage caused by Alzheimer's disease genes, specifically APP (amyloid precursor protein). Humanin acts as a ligand for formyl peptide receptor-like 1 (FPRL1) with APP and utilizes its neuroprotective effects by inhibiting FPRL1 access to APP. The peptide prevents Bax translocation from cytosol to mitochondria. Humanin expression levels may be dependent on defects in energy production in muscles with mitochondrial abnormalities. The peptide has been detected in muscles of patients with the mitochondrial disease chronic progressive external ophthalmoplegia (CPEO). Humanin is mainly expressed in the kidney, heart, liver, testis and skeletal muscles.

Type: Primary
Antigen: Humanin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-24/24
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge