Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ALS2CR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11649R-FITC
BOSSBS-11649R-FITCEA 900 EUR InStock
BOSSBS-11649R-FITC
Anti-ALS2CR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The NGG1 interacting factor 3-like 1 (NIF3L1) is a 377 amino acid protein expressed mainly in the cytoplasm of cells in several different tissues. It has been highly conserved throughout evolution, from bacteria to mammals. NIF3L participates in retinoic acid-primed neural differentiation of P19 embryonic carcinoma cells by cooperating with Trip15/CSN2, a transcriptional corepressor/component of COP9 signalosome. NIF3L1 interacts with itself and with the NIF3L1 binding protein 1 (NIF3L1 BP1), which is a novel protein presumed to contain a leucine zipper domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALS2CR1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge