Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GRC5/PHF2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11659R-CY3
BOSSBS-11659R-CY3EA 1350 EUR InStock
BOSSBS-11659R-CY3
Anti-GRC5/PHF2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
PHF2 is a 1,101 amino acid protein belonging to the PHD finger protein family. Members of the PHD finger protein family function as transcriptional regulators that affect gene expression by modulating chromatin structure. Expressed in a wide variety of tissues, PHF2 localizes to the nucleus and contains one PHD-type zinc finger and one JMJC domain, suggesting a possible role for PHF2 in transcription regulation and chromatin remodeling. The gene encoding PHF2 lies in the candidate region for hereditary sensory neuropathy type I (HSN1), a disorder characterized by sensory dysfunction. PHF2 exhists as two isoforms produced by alternative splicing, designated isoform 1 and 2 respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRC5/PHF2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge