Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GATB Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11667R
BOSSBS-11667REA 725 EUR InStock
BOSSBS-11667R
Anti-GATB Rabbit Polyclonal Antibody
Antikörper
PET112L is the human homolog of the S. cerevisiae COX assembly protein pet112, a protein that is believed to play an important role in the translation of mitochondrial genes. PET112L, also known as HSPC199 or Glu-ADT subunit B (glutamyl-tRNA(Gln) amidotransferase subunit B), is a 557 amino acid protein belonging to the gatB/gatE family of proteins (GatB subfamily) and is believed to play a role in energy metabolism. Localizing to mitochondria, PET112L is expressed in tissues such as heart and muscle, which exhibit high rates of oxidative phosphorylation. The gene encoding PET112L is overexpressed in recurrent ependymoma.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PET112L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge